Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135802.1 | NP_001129274.1 | 15 | 1994 | ||
XM_006251794.2 | XP_006251856.1 | 15 | 2067 | |||
XM_008770506.1 | XP_008768728.1 | 15 | 2060 | |||
GenBank mRNA | BC162031.1 | - | 15 | 1994 | 9101206 |