Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134990.1 | NP_001128462.1 | 2 | 227 | ||
XM_006252059.2 | XP_006252121.1 | 3 | 1080 | |||
XM_006252061.2 | XP_006252123.1 | 2 | 908 | |||
GenBank mRNA | BC168660.1 | - | 2 | 227 | 5621479 | |
FQ230419.1 | - | 3 | 277 |