Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256850.1 | NP_001243779.1 | 21 | 3697 | ||
NM_001267550.2 | NP_001254479.2 | 21 | 3697 | |||
NM_003319.4 | NP_003310.4 | 20 | 3559 | |||
NM_133378.4 | NP_596869.4 | 21 | 3697 | |||
NM_133379.4 | NP_596870.2 | 21 | 3697 | |||
NM_133432.3 | NP_597676.3 | 20 | 3559 | |||
NM_133437.4 | NP_597681.4 | 20 | 3559 | |||
XM_017004819.1 | XP_016860308.1 | 21 | 3700 | |||
XM_017004820.1 | XP_016860309.1 | 21 | 3700 | |||
XM_017004821.1 | XP_016860310.1 | 21 | 3697 | |||
XM_017004822.1 | XP_016860311.1 | 21 | 3700 | |||
XM_017004823.1 | XP_016860312.1 | 21 | 3700 | |||
GenBank mRNA | X90568.1 | - | 20 | 3466 |