Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289395.1 | NP_001276324.1 | 5 | 430 | ||
NM_003322.4 | NP_003313.3 | 6 | 589 | |||
GenBank mRNA | AB593123.1 | - | 6 | 589 | ||
AB593124.1 | - | 6 | 575 | |||
BC032714.1 | - | 5 | 415 | 5495136 | ||
BC065261.1 | - | 5 | 398 | 6144003 | ||
CN355966.1 | - | 2 | 398 | |||
U82468.1 | - | 6 | 581 |