Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169102.1 | NP_001162573.1 | 2 | 104 | ||
XM_006253023.2 | XP_006253085.1 | 2 | 200 | |||
GenBank mRNA | CF109688.1 | - | 2 | 139 |