Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167.3 | NP_001158.2 | 2 | 866 | ||
NM_001204401.1 | NP_001191330.1 | 2 | 833 | |||
XM_006724754.2 | XP_006724817.1 | 3 | 946 | |||
XM_011531329.2 | XP_011529631.1 | 2 | 836 | |||
GenBank mRNA | AK313883.1 | - | 2 | 833 | ||
BC032729.1 | - | 2 | 785 | 5532247 | ||
U32974.1 | - | 2 | 788 |