Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106158.1 | NP_001099628.1 | 10, 11 | 1598 | ||
XM_008772030.1 | XP_008770252.1 | 9, 10 | 1222 | |||
XM_008772031.1 | XP_008770253.1 | 9, 10 | 1222 | |||
XM_008772032.1 | XP_008770254.1 | 9, 10 | 1222 | |||
XM_008772033.1 | XP_008770255.1 | 9, 10 | 1222 | |||
GenBank mRNA | FQ228342.1 | - | 8, 9 | 1110 |