Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 4 | 328 | ||
NM_005659.6 | NP_005650.2 | 4 | 328 | |||
XM_011530361.2 | XP_011528663.1 | 4 | 454 | |||
GenBank mRNA | AF141201.1 | - | 3 | 198 | ||
AJ239058.1 | - | 3 | 198 | |||
AK225877.1 | - | 4 | 285 | |||
AK304734.1 | - | 4 | 298 | |||
AK308786.1 | - | 4 | 272 | |||
AK310950.1 | - | 4 | 280 | |||
AK315998.1 | - | 4 | 279 | |||
AY101594.1 | - | 4 | 255 | |||
BC001049.2 | - | 4 | 275 | 3507963 | ||
BC005087.1 | - | 4 | 276 | 3830774 | ||
BM761958.1 | - | 3 | 267 | |||
BX648483.1 | - | 4 | 260 | |||
CB128364.1 | - | 4 | 328 | |||
CR456607.1 | - | 4 | 212 | |||
DB496066.1 | - | 4 | 312 | |||
U64444.1 | - | 4 | 276 |