Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001074.2 | NP_001065.2 | 1 | 398 | ||
XM_011532229.2 | XP_011530531.1 | 1 | 399 | |||
XM_011532230.1 | XP_011530532.1 | 1 | 399 | |||
GenBank mRNA | AK223142.1 | - | 1 | 372 | ||
AK313190.1 | - | 1 | 399 | |||
BC030974.1 | - | 1 | 398 | 4593426 | ||
J05428.1 | - | 1 | 366 | |||
S82485.1 | - | 1 | 76 |