Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075.5 | NP_001066.1 | 1 | 349 | ||
NM_001144767.2 | NP_001138239.1 | 1 | 349 | |||
XM_017008585.1 | XP_016864074.1 | 1 | 361 | |||
GenBank mRNA | AK222872.1 | - | 1 | 351 | ||
AK292738.1 | - | 1 | 355 | |||
AK298432.1 | - | 1 | 348 | |||
BC113649.1 | - | 1 | 350 | 8322701 |