Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076.3 | NP_001067.2 | 1 | 593 | ||
GenBank mRNA | AF180322.1 | - | 1 | 473 | ||
AF548389.1 | - | 1 | 452 | |||
AK289419.1 | - | 1 | 466 | |||
BC171879.1 | - | 593 | 9054533 | |||
U08854.1 | - | 1 | 456 |