Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106239.1 | NP_001099709.1 | 3 | 248 | ||
XM_006228651.1 | XP_006228713.1 | 3 | 324 | |||
XM_006228652.2 | XP_006228714.1 | 3 | 297 |