Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014810.1 | NP_001014810.1 | 6 | 671 | ||
NM_198734.2 | NP_942029.2 | 6 | 681 | |||
XM_008759128.1 | XP_008757350.1 | 6 | 700 | |||
XM_008759129.1 | XP_008757351.1 | 6 | 680 | |||
GenBank mRNA | CB576820.1 | - | 6 | 624 | ||
CB751111.1 | - | 3 | 277 | |||
CB809766.1 | - | 1 | 15 |
Gene Symbol: LOC103689986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228658.2 | XP_006228720.1 | 6 | 700 | ||
XM_006228660.2 | XP_006228722.1 | 6 | 680 | |||
XM_008759221.1 | XP_008757443.1 | 6 | 710 | |||
XM_008759222.1 | XP_008757444.1 | 6 | 729 |