Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 3, 4 | 1089 | ||
NR_033434.1 | 2, 3 | 935 | ||||
NR_033437.1 | 4, 5 | 1188 | ||||
GenBank mRNA | AB062285.1 | - | 3, 4 | 1011 | ||
AK291306.1 | - | 3, 4 | 1005 | |||
AL080099.2 | - | 2, 3 | 868 | |||
BC000364.2 | - | 3, 4 | 1006 | 2964556 | ||
BC007511.2 | - | 3, 4 | 1006 | 2964556 | ||
CR456787.1 | - | 3, 4 | 983 | |||
D86227.1 | - | 3, 4 | 1002 | |||
D86228.1 | - | 3, 4 | 1002 | |||
D86230.1 | - | 3, 4 | 1002 | |||
EU921886.1 | - | 3, 4 | 1003 | |||
EU921887.1 | - | 2, 3 | 849 | |||
EU921888.1 | - | 2, 3 | 849 | |||
EU921889.1 | - | 3, 4 | 948 | |||
EU921891.1 | - | 4, 5 | 1102 | |||
EU921893.1 | - | 3, 4 | 656 | |||
EU921894.1 | - | 3, 4 | 1003 | |||
J03626.1 | - | 3, 4 | 1087 | |||
KU178375.1 | - | 3, 4 | 983 | |||
KU178376.1 | - | 4, 5 | 1082 | |||
M36661.1 | - | 3, 4 | 949 |