Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000120.1 | NP_001000120.1 | 1 | 839 |
Gene Symbol: Olr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000689.1 | NP_001000689.1 | 1 | 839 |