Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001089591.1 | NP_001083060.1 | 1 | 147 |
Gene Symbol: UQCRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297565.1 | NP_001284494.1 | 4 | 399 | ||
NM_001297566.1 | NP_001284495.1 | 2 | 224 | |||
NM_006004.3 | NP_005995.2 | 3 | 251 | |||
GenBank mRNA | AK311967.1 | - | 3 | 147 | ||
BC001426.2 | - | 3 | 151 | 3140323 | ||
BC001934.1 | - | 3 | 156 | 2823237 | ||
BC015177.1 | - | 3 | 154 | 3905169 | ||
BC093060.1 | - | 4 | 314 | 30409772 | ||
BC107703.1 | - | 3 | 157 | 6605146 | ||
BG472687.1 | - | 2 | 125 | |||
BP366403.1 | - | 4 | 401 | |||
BT007070.1 | - | 3 | 97 | |||
M36647.1 | - | 3 | 127 | |||
Y00764.1 | - | 3 | 133 |