Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000130.1 | NP_001000130.1 | 1 | 426 |
Gene Symbol: LOC103690133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759842.1 | XP_008758064.1 | 1 | 108 |