Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDM6A SIRNA Assays ›
Gene Symbol:
KDM6A
Gene Name:
lysine demethylase 6A
Gene Aliases:
KABUK2, UTX, bA386N14.2
Chromosome Location:
Chr.X: 44873175 - 45112612 on Build GRCh38
UniGene ID:
Hs.522616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14736
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KDM6A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291415.1
NP_001278344.1
21
3604
NM_001291416.1
NP_001278345.1
20
3469
NM_001291417.1
NP_001278346.1
19
3313
NM_001291418.1
NP_001278347.1
18
3211
NM_001291421.1
NP_001278350.1
18
3192
NM_021140.3
NP_066963.2
20
3448
NR_111960.1
19
3367
XM_005272656.4
XP_005272713.1
20
3502
XM_005272659.4
XP_005272716.1
19
3346
XM_011543957.2
XP_011542259.1
22
3661
XM_011543958.2
XP_011542260.1
21
3604
XM_011543959.2
XP_011542261.1
21
3559
XM_011543960.2
XP_011542262.1
22
3661
XM_011543961.2
XP_011542263.1
21
3526
XM_011543962.2
XP_011542264.1
21
3505
XM_011543963.2
XP_011542265.1
20
3469
XM_011543964.2
XP_011542266.1
20
3448
XM_011543965.2
XP_011542267.1
20
3424
XM_011543966.2
XP_011542268.1
20
3403
XM_011543967.2
XP_011542269.1
21
3505
XM_011543968.2
XP_011542270.1
20
3370
XM_011543969.2
XP_011542271.1
19
3367
XM_011543970.2
XP_011542272.1
19
3346
XM_011543971.2
XP_011542273.1
20
3424
XM_011543972.2
XP_011542274.1
19
3313
XM_011543973.2
XP_011542275.1
20
3370
XM_011543974.1
XP_011542276.1
20
3448
XM_011543975.2
XP_011542277.1
17
2774
XM_017029782.1
XP_016885271.1
21
3526
XM_017029783.1
XP_016885272.1
19
3367
XM_017029784.1
XP_016885273.1
19
2914
XM_017029785.1
XP_016885274.1
17
2887
GenBank mRNA
AB208795.1
-
20
3401
AF000992.1
-
20
3097
AF000993.1
-
20
3097
AK294456.1
-
20
3044
AK303430.1
-
9
2052
AK304114.1
-
19
3382
AK307691.1
-
9
2081
AL831996.2
-
9
1974
BC093868.1
-
20
3094
7939713
BC113381.1
-
20
3094
8322433
BC143272.1
-
20
3115
9051774
BC143277.1
-
21
3250
9051779
BC143278.1
-
19
2959
9051780
BX640694.1
-
8
1853
DQ062677.1
-
19
2834
DQ062678.1
-
19
2936
EU026283.1
-
20
3092
EU026291.1
-
19
2990
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
neural tube closure
heart morphogenesis
respiratory system process
positive regulation of gene expression
somite rostral/caudal axis specification
multicellular organism growth
mesodermal cell differentiation
notochord morphogenesis
histone H3-K4 methylation
oxidation-reduction process
canonical Wnt signaling pathway
histone H3-K27 demethylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
chromatin DNA binding
histone demethylase activity
identical protein binding
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0