Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003370.3 | NP_003361.1 | 2 | 454 | ||
XM_005259199.1 | XP_005259256.1 | 2 | 442 | |||
XM_005259200.1 | XP_005259257.1 | 2 | 442 | |||
XM_017027200.1 | XP_016882689.1 | 2 | 442 | |||
GenBank mRNA | AK314812.1 | - | 2 | 421 | ||
AL556269.3 | - | 2 | 378 | |||
BC026019.1 | - | 2 | 394 | 3345963 | ||
BC038224.1 | - | 2 | 372 | 5210128 | ||
BG751094.1 | - | 2 | 451 | |||
BG825983.1 | - | 2 | 456 | |||
KU178381.1 | - | 1 | 112 | |||
KU178382.1 | - | 1 | 112 | |||
KU178383.1 | - | 1 | 112 | |||
KU178384.1 | - | 1 | 112 | |||
KU178385.1 | - | 1 | 112 | |||
Z46389.1 | - | 2 | 366 |