Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100519.1 | NP_001093989.1 | 8, 9 | 887 | ||
GenBank mRNA | BC098781.1 | - | 2, 3 | 165 | ||
CO558907.1 | - | 282 |
Gene Symbol: LOC103694867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773816.1 | XP_008772038.1 | 7, 8 | 1393 |