Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011935.2 | NP_001011935.2 | 13 | 2145 | ||
XM_008759870.1 | XP_008758092.1 | 13 | 2247 | |||
GenBank mRNA | CA511395.1 | - | 4 | 327 |