Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 21 | 3232 | ||
NM_014000.2 | NP_054706.1 | 22 | 3436 | |||
GenBank mRNA | AK126978.1 | - | 21 | 3486 | ||
AK296509.1 | - | 10 | 1503 | |||
AK300280.1 | - | 7 | 1013 | |||
AL832130.1 | - | 15 | 2358 | |||
BC039174.1 | - | 21 | 3219 | 4520338 | ||
BX537994.1 | - | 23 | 5942 | |||
EU794673.1 | - | 22 | 3436 | |||
M33308.1 | - | 21 | 3188 |