Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106334.1 | NP_001099804.1 | 13 | 1523 | ||
XM_006230976.2 | XP_006231038.1 | 13 | 1548 | |||
XM_006230977.2 | XP_006231039.1 | 12 | 1481 |