Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037349.2 | NP_001032426.1 | 3, 4 | 306 | ||
XM_008760400.1 | XP_008758622.1 | 3, 4 | 333 | |||
XM_008760401.1 | XP_008758623.1 | 4, 5 | 408 | |||
GenBank mRNA | AY786315.1 | - | 3, 4 | 287 | ||
EF061226.2 | - | 4, 5 | 362 |