Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c6v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013904.1 | NP_001013926.1 | 6 | 868 | ||
XM_008759612.1 | XP_008757834.1 | 7 | 901 | |||
GenBank mRNA | BC089886.1 | - | 6 | 868 | 7370532 | |
BC100092.1 | - | 4 | 495 | |||
K03501.1 | - | 843 | ||||
M13711.1 | - | 6 | 843 | |||
M18336.1 | - | 6 | 878 |