Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106361.1 | NP_001099831.1 | 8 | 954 | ||
XM_006231443.2 | XP_006231505.1 | 9 | 1672 | |||
XM_008760403.1 | XP_008758625.1 | 9 | 1669 | |||
XM_008760404.1 | XP_008758626.1 | 8 | 1603 | |||
XM_008760405.1 | XP_008758627.1 | 8 | 1600 |