Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047856.2 | NP_001041321.2 | 3 | 614 | ||
GenBank mRNA | AY383685.1 | - | 535 | |||
FM118556.1 | - | 3 | 614 | |||
FQ214391.1 | - | 3 | 600 | |||
FQ220041.1 | - | 1 | 47 |