Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000277.1 | NP_001000277.1 | 1 | 494 |
Gene Symbol: Olr1726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000896.1 | NP_001000896.1 | 1 | 494 |
Gene Symbol: Olr1739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001423.1 | NP_001001423.1 | 1 | 494 |