Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003386.2 | NP_003377.2 | 28 | 5390 | ||
NM_173059.2 | NP_775082.2 | 28 | 5390 | |||
NR_111917.1 | 28 | 5390 | ||||
NR_111918.1 | 28 | 5390 | ||||
NR_111919.1 | 28 | 5390 | ||||
XM_011516555.2 | XP_011514857.1 | 28 | 5123 | |||
XM_017012581.1 | XP_016868070.1 | 15 | 2879 | |||
GenBank mRNA | AF332975.1 | - | 28 | 5390 | ||
AF332976.1 | - | 28 | 5390 | |||
AF332977.1 | - | 28 | 5390 | |||
AF332978.1 | - | 28 | 5390 | |||
AF332979.1 | - | 28 | 5390 | |||
AF332980.1 | - | 28 | 5390 | |||
AK302596.1 | - | 7 | 1018 |