Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212495.1 | NP_997660.1 | 3 | 1116 | ||
XM_006256097.2 | XP_006256159.1 | 4 | 1789 | |||
XM_008772719.1 | XP_008770941.1 | 3 | 486 | |||
GenBank mRNA | FQ226688.1 | - | 1 | 96 |
Gene Symbol: LOC100909544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256037.2 | XP_006256099.1 | 3 | 1695 | ||
XM_008772772.1 | XP_008770994.1 | 3 | 485 | |||
XM_008772773.1 | XP_008770995.1 | 4 | 1787 |