Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145853.1 | NP_001139325.1 | 8 | 1184 | ||
NM_006005.3 | NP_005996.2 | 8 | 1188 | |||
XM_017008586.1 | XP_016864075.1 | 9 | 1167 | |||
GenBank mRNA | AF084481.1 | - | 8 | 1188 | ||
BC030130.2 | - | 8 | 1118 | 4127654 | ||
KU178395.1 | - | 7 | 1018 | |||
Y18064.1 | - | 8 | 1176 |