Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106419.1 | NP_001099889.1 | 3 | 554 | ||
XM_006232058.2 | XP_006232120.1 | 3 | 472 | |||
XM_006232059.2 | XP_006232121.1 | 3 | 436 |