Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760900.1 | XP_008759122.1 | 2 | 561 | ||
XM_008760901.1 | XP_008759123.1 | 2 | 524 |