Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761239.1 | XP_008759461.1 | 9, 10 | 1542 | ||
XM_008761240.1 | XP_008759462.1 | 9, 10 | 1542 | |||
XM_008761241.1 | XP_008759463.1 | 9, 10 | 1467 | |||
XM_008761242.1 | XP_008759464.1 | 9, 10 | 1434 |