Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106446.1 | NP_001099916.1 | 2 | 1009 | ||
XM_006232614.2 | XP_006232676.1 | 4 | 1391 | |||
XM_008761136.1 | XP_008759358.1 | 3 | 1516 | |||
XM_008761137.1 | XP_008759359.1 | 4 | 1410 |