Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106470.1 | NP_001099940.1 | 10 | 1040 | ||
XM_006233266.2 | XP_006233328.1 | 10 | 1100 | |||
GenBank mRNA | BC166789.1 | - | 10 | 946 | 7936402 |