Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167604.1 | NP_001161076.1 | 19 | 1866 | ||
NM_001324128.1 | NP_001311057.1 | 18 | 1785 | |||
NM_001324131.1 | NP_001311060.1 | 20 | 1982 | |||
NM_001324132.1 | NP_001311061.1 | 19 | 1849 | |||
NM_001324133.1 | NP_001311062.1 | 20 | 1938 | |||
NM_001324134.1 | NP_001311063.1 | 18 | 1769 | |||
NM_001324135.1 | NP_001311064.1 | 19 | 1887 | |||
NM_001324136.1 | NP_001311065.1 | 20 | 2153 | |||
NM_020383.3 | NP_065116.3 | 20 | 1938 | |||
XM_011540134.1 | XP_011538436.1 | 19 | 1758 | |||
XM_017016613.1 | XP_016872102.1 | 19 | 1788 | |||
XM_017016614.1 | XP_016872103.1 | 21 | 1921 | |||
XM_017016615.1 | XP_016872104.1 | 19 | 1805 | |||
GenBank mRNA | AF195530.1 | - | 19 | 1765 | ||
AF272981.1 | - | 18 | 1689 | |||
AK093693.1 | - | 18 | 1743 | |||
AK095447.1 | - | 5 | 1984 | |||
AK223513.1 | - | 19 | 1749 | |||
AK289436.1 | - | 20 | 1841 | |||
AL833411.1 | - | 19 | 1772 | |||
BC005126.2 | - | 19 | 1727 | 3836436 | ||
BC007579.1 | - | 20 | 1829 | 3139868 | ||
BC013417.2 | - | 16 | 1439 | |||
BX537682.1 | - | 16 | 3783 | |||
CR456922.1 | - | 18 | 1689 | |||
X95762.1 | - | 18 | 1689 |