Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 21 | 3390 | ||
XM_005264544.1 | XP_005264601.1 | 20 | 3367 | |||
XM_005264546.1 | XP_005264603.1 | 19 | 2498 | |||
XM_006712094.2 | XP_006712157.1 | 22 | 3533 | |||
XM_011533097.1 | XP_011531399.1 | 21 | 2699 | |||
XM_011533098.1 | XP_011531400.1 | 20 | 3232 | |||
XM_011533099.2 | XP_011531401.1 | 21 | 4340 | |||
GenBank mRNA | AK094339.1 | - | 3 | 2194 | ||
AK124769.1 | - | 10 | 2267 | |||
AK289920.1 | - | 21 | 3069 | |||
AK299039.1 | - | 12 | 1792 | |||
AK312426.1 | - | 21 | 2827 | |||
AK316481.1 | - | 12 | 1806 | |||
AL833550.1 | - | 20 | 5335 | |||
BC032847.2 | - | 22 | 2791 | 5267242 | ||
BX647758.1 | - | 21 | 3372 | |||
CR749840.1 | - | 22 | 3164 | |||
D89729.1 | - | 20 | 2680 | |||
Y08614.2 | - | 21 | 2700 |