Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100527.1 | NP_001093997.1 | 5 | 1085 | ||
XM_006234429.2 | XP_006234491.1 | 4 | 1509 | |||
GenBank mRNA | BC098777.1 | - | 2 | 183 | ||
BC166428.1 | - | 5 | 1085 | 7106784 |
Set Membership: |
G-protein coupled receptor |