Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000350.1 | NP_001000350.1 | 1 | 692 |
Gene Symbol: Olr674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000352.1 | NP_001000352.1 | 1 | 692 |