Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079.3 | NP_001070.2 | 11 | 1579 | ||
NM_207519.1 | NP_997402.1 | 3 | 597 | |||
XM_017004867.1 | XP_016860356.1 | 13 | 2980 | |||
XM_017004868.1 | XP_016860357.1 | 13 | 2962 | |||
XM_017004869.1 | XP_016860358.1 | 13 | 2980 | |||
XM_017004870.1 | XP_016860359.1 | 13 | 2980 | |||
GenBank mRNA | AB083211.1 | - | 3 | 597 | ||
AL832287.1 | - | 4 | 2424 | |||
BC039039.1 | - | 10 | 1233 | 5168073 | ||
BC053878.1 | - | 11 | 1555 | 5757161 | ||
L05148.1 | - | 11 | 1581 |