Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106506.1 | NP_001099976.1 | 12, 13 | 2000 | ||
XM_006234961.2 | XP_006235023.1 | 13, 14 | 2244 | |||
XM_006234963.2 | XP_006235025.1 | 13, 14 | 2241 | |||
XM_006234964.2 | XP_006235026.1 | 13, 14 | 2241 | |||
XM_006234965.2 | XP_006235027.1 | 13, 14 | 2238 | |||
GenBank mRNA | BC168839.1 | - | 8, 9 | 964 |