Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000484.3 | NP_000475.1 | 2 | 327 | ||
NM_001136016.3 | NP_001129488.1 | 2 | 308 | |||
NM_001136131.2 | NP_001129603.1 | 2 | 214 | |||
NM_001204301.1 | NP_001191230.1 | 2 | 327 | |||
NM_001204302.1 | NP_001191231.1 | 2 | 327 | |||
NM_001204303.1 | NP_001191232.1 | 2 | 327 | |||
NM_201413.2 | NP_958816.1 | 2 | 327 | |||
NM_201414.2 | NP_958817.1 | 2 | 327 | |||
GenBank mRNA | A02759.1 | - | 2 | 273 | ||
AK295621.1 | - | 2 | 269 | |||
AK296229.1 | - | 2 | 214 | |||
AK297229.1 | - | 2 | 191 | |||
AK297412.1 | - | 2 | 275 | |||
AK311717.1 | - | 2 | 387 | |||
AK312326.1 | - | 2 | 192 | |||
BC004369.1 | - | 2 | 253 | |||
BC065529.1 | - | 2 | 258 | 6152423 | ||
BI559391.1 | - | 2 | 332 | |||
BM172428.1 | - | 2 | 327 | |||
X06989.1 | - | 2 | 251 | |||
Y00264.1 | - | 2 | 274 |