Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004231.1 | NP_001004231.1 | 4, 5 | 632 | ||
XM_006233584.2 | XP_006233646.1 | 3, 4 | 1270 | |||
GenBank mRNA | BC079156.1 | - | 4, 5 | 632 | 7128989 | |
FQ214366.1 | - | 4, 5 | 476 |