Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011962.1 | NP_001011962.1 | 6, 7 | 605 | ||
XM_006233768.2 | XP_006233830.1 | 3, 4 | 1273 | |||
XM_006233769.2 | XP_006233831.1 | 4, 5 | 665 | |||
XM_006233770.2 | XP_006233832.1 | 6, 7 | 1645 | |||
GenBank mRNA | BC085782.1 | - | 6, 7 | 605 | 7107449 |