Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 1164 | ||
NM_001256280.1 | NP_001243209.1 | 3 | 969 | |||
NM_019591.3 | NP_062537.2 | 4 | 1065 | |||
XM_005266182.3 | XP_005266239.1 | 5 | 1152 | |||
XM_005266183.3 | XP_005266240.1 | 6 | 1355 | |||
XM_011534829.1 | XP_011533131.1 | 5 | 1268 | |||
XM_017019920.1 | XP_016875409.1 | 6 | 1788 | |||
XM_017019921.1 | XP_016875410.1 | 6 | 1251 | |||
XM_017019922.1 | XP_016875411.1 | 5 | 2137 | |||
XM_017019923.1 | XP_016875412.1 | 5 | 2068 | |||
GenBank mRNA | AK000762.1 | - | 1 | 268 | ||
AK122941.1 | - | 3 | 3178 | |||
AK225296.1 | - | 1 | 268 | |||
AK291243.1 | - | 4 | 1032 | |||
BC046206.2 | - | 4 | 966 | 6176427 | ||
EU668356.1 | - | 4 | 943 |