Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 1173 | ||
NR_036599.1 | 5 | 1291 | ||||
NR_036600.1 | 3 | 1046 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 1148 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 1203 | |||
GenBank mRNA | AK131368.1 | - | 2 | 1771 | ||
AK131574.1 | - | 1 | 558 | |||
AK293074.1 | - | 3 | 1046 | |||
AK304704.1 | - | 4 | 1137 | |||
BC140720.1 | - | 3 | 1074 | 9021588 | ||
BX640767.1 | - | 1 | 200 | |||
CR933598.1 | - | 4 | 1136 |