Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039686.1 | NP_001034775.1 | 3, 4 | 234 | ||
XM_008763064.1 | XP_008761286.1 | 2, 3 | 1762 | |||
XM_008763065.1 | XP_008761287.1 | 6, 7 | 718 | |||
GenBank mRNA | AY943907.1 | - | 3, 4 | 234 |