Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005368.2 | NP_001005368.1 | 3 | 856 | ||
NM_001324164.1 | NP_001311093.1 | 3 | 824 | |||
NM_001324165.1 | NP_001311094.1 | 3 | 962 | |||
NM_001324166.1 | NP_001311095.1 | 3 | 726 | |||
NM_001324167.1 | NP_001311096.1 | 3 | 722 | |||
NM_001324168.1 | NP_001311097.1 | 3 | 860 | |||
NM_006973.2 | NP_008904.1 | 3 | 718 | |||
XM_017016616.1 | XP_016872105.1 | 2 | 3789 | |||
GenBank mRNA | AK312132.1 | - | 3 | 715 | ||
BC022842.1 | - | 2 | 653 | 5014207 | ||
BG477527.1 | - | 3 | 728 | |||
BM127643.1 | - | 1 | 259 | |||
U69645.1 | - | 3 | 694 |