Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106610.2 | NP_001100080.2 | 9, 10 | 905 | ||
GenBank mRNA | BC166430.1 | - | 9, 10 | 905 | 7131565 |